Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF821 SIRNA Assays ›
Gene Symbol:
ZNF821
Gene Name:
zinc finger protein 821
Gene Aliases:
-
Chromosome Location:
Chr.16: 71859680 - 71886224 on Build GRCh38
UniGene ID:
Hs.744055
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s30988
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF821
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001201552.1
NP_001188481.1
8
1019
NM_001201553.1
NP_001188482.1
7
1004
NM_001201554.1
NP_001188483.1
7
893
NM_001201556.1
NP_001188485.1
7
852
NM_001318238.1
NP_001305167.1
8
973
NM_001318239.1
NP_001305168.1
7
746
NM_017530.2
NP_060000.1
6
878
XM_005256033.2
XP_005256090.1
9
1093
XM_006721233.1
XP_006721296.1
8
952
XM_011523211.2
XP_011521513.1
7
798
XM_011523212.2
XP_011521514.1
7
851
XM_011523214.2
XP_011521516.1
7
821
XM_011523215.2
XP_011521517.1
5
492
XM_011523220.2
XP_011521522.1
6
574
XM_017023409.1
XP_016878898.1
8
937
XM_017023410.1
XP_016878899.1
8
1684
XM_017023411.1
XP_016878900.1
8
761
XM_017023412.1
XP_016878901.1
9
1093
XM_017023413.1
XP_016878902.1
7
821
XM_017023414.1
XP_016878903.1
7
880
XM_017023415.1
XP_016878904.1
7
818
XM_017023416.1
XP_016878905.1
8
1030
XM_017023417.1
XP_016878906.1
8
1024
XM_017023418.1
XP_016878907.1
8
886
XM_017023419.1
XP_016878908.1
8
868
XM_017023420.1
XP_016878909.1
9
1113
XM_017023421.1
XP_016878910.1
7
879
XM_017023422.1
XP_016878911.1
6
534
XM_017023423.1
XP_016878912.1
7
860
XM_017023424.1
XP_016878913.1
6
714
XM_017023425.1
XP_016878914.1
5
568
GenBank mRNA
AF070588.1
-
6
846
AK296603.1
-
7
1004
AK313693.1
-
7
853
AL538838.3
-
7
871
BC012116.1
-
7
850
4553333
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, sequence-specific DNA binding
protein binding
transcription regulatory region DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline