Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017539.2 | NP_060009.1 | 12 | 1730 | ||
XM_006721062.1 | XP_006721125.1 | 12 | 1826 | |||
XM_011545882.2 | XP_011544184.1 | 13 | 2266 | |||
XM_011545883.1 | XP_011544185.1 | 13 | 2034 | |||
XM_011545885.2 | XP_011544187.1 | 13 | 2266 | |||
XM_011545886.2 | XP_011544188.1 | 5 | 641 | |||
XM_017023426.1 | XP_016878915.1 | 13 | 2266 | |||
XM_017023428.1 | XP_016878917.1 | 13 | 2266 | |||
XM_017023429.1 | XP_016878918.1 | 13 | 2267 | |||
XM_017023431.1 | XP_016878920.1 | 13 | 2267 | |||
XM_017023432.1 | XP_016878921.1 | 12 | 1826 | |||
GenBank mRNA | AF494040.1 | - | 12 | 1730 | ||
AK056509.2 | - | 12 | 1826 | |||
AK303958.1 | - | 12 | 1631 |