Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017539.2 | NP_060009.1 | 34 | 4796 | ||
XM_006721062.1 | XP_006721125.1 | 34 | 4934 | |||
XM_011545882.2 | XP_011544184.1 | 35 | 5374 | |||
XM_011545883.1 | XP_011544185.1 | 35 | 5142 | |||
XM_011545885.2 | XP_011544187.1 | 35 | 5374 | |||
XM_011545886.2 | XP_011544188.1 | 27 | 3749 | |||
XM_011545888.2 | XP_011544190.1 | 17 | 2193 | |||
XM_011545889.2 | XP_011544191.1 | 17 | 2157 | |||
XM_017023426.1 | XP_016878915.1 | 35 | 5374 | |||
XM_017023427.1 | XP_016878916.1 | 33 | 5131 | |||
XM_017023428.1 | XP_016878917.1 | 35 | 5374 | |||
XM_017023429.1 | XP_016878918.1 | 35 | 5375 | |||
XM_017023431.1 | XP_016878920.1 | 35 | 5375 | |||
XM_017023432.1 | XP_016878921.1 | 34 | 4934 | |||
GenBank mRNA | AF494040.1 | - | 34 | 4796 | ||
AK125907.1 | - | 11 | 1513 | |||
BC019878.1 | - | 1 | 78 | |||
EU668352.1 | - | 1 | 78 |