Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198321.3 | NP_938080.1 | 11, 12 | 1780 | ||
GenBank mRNA | AB078145.1 | - | 11, 12 | 1648 | ||
AJ505950.1 | - | 11, 12 | 1648 | |||
AK023782.1 | - | 8, 9 | 1114 | |||
AK023815.1 | - | 2, 3 | 230 | |||
AK127135.1 | - | 11, 12 | 1749 | |||
AL096739.1 | - | 4, 5 | 924 | |||
BC007224.1 | - | 4, 5 | 580 | |||
BC072450.1 | - | 7, 8 | 1080 | |||
BC098444.1 | - | 6, 7 | 769 |