Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198321.3 | NP_938080.1 | 9 | 1330 | ||
GenBank mRNA | AB078145.1 | - | 9 | 1198 | ||
AJ505950.1 | - | 9 | 1198 | |||
AK023782.1 | - | 6 | 664 | |||
AK127135.1 | - | 9 | 1299 | |||
AL096739.1 | - | 2 | 474 | |||
BC007224.1 | - | 2 | 130 | |||
BC072450.1 | - | 5 | 630 | |||
BC098444.1 | - | 4 | 319 |