Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNOT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017546.4 | NP_060016.3 | 5 | 1152 | ||
GenBank mRNA | AF103798.1 | - | 5 | 1143 | ||
AK024221.1 | - | 4 | 569 | |||
BC018664.2 | - | 5 | 1061 | |||
BC064421.1 | - | 5 | 1119 | 5752134 |