Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134422.1 | NP_001127894.1 | 5 | 1926 | ||
NM_001134423.2 | NP_001127895.1 | 5 | 1387 | |||
NM_001282762.1 | NP_001269691.1 | 5 | 1085 | |||
NM_001282763.1 | NP_001269692.1 | 5 | 612 | |||
NM_001282765.1 | NP_001269694.1 | 5 | 1545 | |||
NM_017548.4 | NP_060018.1 | 5 | 1189 | |||
XM_005247588.4 | XP_005247645.1 | 6 | 1219 | |||
XM_005247589.4 | XP_005247646.1 | 6 | 1216 | |||
XM_005247590.4 | XP_005247647.1 | 5 | 1213 | |||
XM_005247592.4 | XP_005247649.1 | 6 | 693 | |||
XM_005247594.4 | XP_005247651.1 | 6 | 597 | |||
XM_011512961.2 | XP_011511263.1 | 5 | 1950 | |||
XM_011512962.2 | XP_011511264.1 | 6 | 521 | |||
XM_017006808.1 | XP_016862297.1 | 5 | 1381 | |||
GenBank mRNA | AF103803.1 | - | 5 | 1057 | ||
AK025647.1 | - | 2 | 208 | |||
AK096865.1 | - | 5 | 1365 | |||
BG773216.1 | - | 5 | 617 |