Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017564.9 | NP_060034.9 | 58 | 6407 | ||
XM_011538537.2 | XP_011536839.1 | 58 | 6448 | |||
XM_011538538.2 | XP_011536840.1 | 58 | 6448 | |||
XM_011538539.2 | XP_011536841.1 | 44 | 4651 | |||
XM_011538541.2 | XP_011536843.1 | 36 | 3758 | |||
XM_011538542.2 | XP_011536844.1 | 25 | 2567 | |||
XM_017019585.1 | XP_016875074.1 | 58 | 6448 | |||
GenBank mRNA | AB052958.1 | - | 58 | 6403 | ||
AF160476.1 | - | 13 | 1429 | |||
AJ295695.1 | - | 58 | 6407 | |||
AK024503.1 | - | 26 | 2749 | |||
AK074051.1 | - | 36 | 3778 | |||
AK160380.1 | - | 57 | 6125 | |||
AL133021.1 | - | 18 | 1889 | |||
AY227444.1 | - | 27 | 2816 | |||
AY311388.1 | - | 58 | 6425 |