Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017581.3 | NP_060051.2 | 2, 3 | 344 | ||
GenBank mRNA | AF227732.1 | - | 2, 3 | 187 | ||
AJ243342.1 | - | 2, 3 | 221 | |||
BC113549.1 | - | 2, 3 | 300 | 8322601 | ||
BC113575.1 | - | 2, 3 | 300 | 8322627 |