Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2Q1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017582.6 | NP_060052.3 | 7 | 923 | ||
GenBank mRNA | AK056388.1 | - | 3 | 223 | ||
AK293986.1 | - | 7 | 546 | |||
AY112698.1 | - | 7 | 898 | |||
AY948200.1 | - | 7 | 906 | |||
BC000848.1 | - | 3 | 257 | |||
BC009286.2 | - | 2 | 61 | |||
BC061583.1 | - | 3 | 235 | |||
BC070158.1 | - | 6 | 448 | |||
BC087836.1 | - | 5 | 334 | |||
BC107057.2 | - | 8 | 591 | |||
BC148185.2 | - | 7 | 600 |