Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2Q1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017582.6 | NP_060052.3 | 2, 3 | 521 | ||
GenBank mRNA | AK293986.1 | - | 2, 3 | 144 | ||
AY112698.1 | - | 2, 3 | 496 | |||
AY948200.1 | - | 2, 3 | 504 | |||
BC070158.1 | - | 1, 2 | 46 | |||
BC107057.2 | - | 3, 4 | 189 | |||
BC148185.2 | - | 2, 3 | 198 | |||
BQ278712.1 | - | 521 |