Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017612.4 | NP_060082.2 | 1, 2 | 435 | ||
XM_006719491.2 | XP_006719554.1 | 1, 2 | 351 | |||
GenBank mRNA | AK027702.1 | - | 1, 2 | 304 | ||
AK074638.1 | - | 1, 2 | 333 | |||
BC065918.1 | - | 1, 2 | 274 | 5752053 | ||
DB048019.1 | - | 1, 2 | 436 | |||
DB456426.1 | - | 1, 2 | 358 |