Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKN2AIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317343.1 | NP_001304272.1 | 3 | 781 | ||
NM_017632.3 | NP_060102.1 | 3 | 812 | |||
GenBank mRNA | AF246705.1 | - | 3 | 760 | ||
AK000043.1 | - | 3 | 767 | |||
AK096180.1 | - | 3 | 648 | |||
AK225848.1 | - | 3 | 625 | |||
BC022270.1 | - | 3 | 649 | 4697255 | ||
BX538162.1 | - | 2 | 285 |