Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARMIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173977.1 | NP_001167448.1 | 28 | 2900 | ||
NM_017640.5 | NP_060110.4 | 28 | 2900 | |||
XM_005249218.1 | XP_005249275.1 | 28 | 2900 | |||
XM_017011008.1 | XP_016866497.1 | 29 | 2912 | |||
XM_017011009.1 | XP_016866498.1 | 29 | 2912 | |||
XM_017011010.1 | XP_016866499.1 | 28 | 2900 | |||
XM_017011011.1 | XP_016866500.1 | 29 | 2912 | |||
XM_017011012.1 | XP_016866501.1 | 29 | 2912 | |||
XM_017011013.1 | XP_016866502.1 | 29 | 2912 | |||
XM_017011014.1 | XP_016866503.1 | 28 | 2852 | |||
XM_017011015.1 | XP_016866504.1 | 29 | 2912 | |||
XM_017011016.1 | XP_016866505.1 | 29 | 2912 | |||
XM_017011017.1 | XP_016866506.1 | 29 | 2912 | |||
GenBank mRNA | AK000055.1 | - | 3 | 237 | ||
AK225208.1 | - | 3 | 237 | |||
BC140770.1 | - | 28 | 2681 | 9021638 | ||
FJ009082.1 | - | 28 | 2532 | |||
Y10508.1 | - | 5 | 519 | |||
Z83950.1 | - | 3 | 301 |