Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017664.3 | NP_060134.2 | 6 | 984 | ||
NR_104587.1 | 7 | 1046 | ||||
GenBank mRNA | AK000100.1 | - | 6 | 946 | ||
AK000111.1 | - | 1 | 277 | |||
AK026017.1 | - | 2 | 58 | |||
AK094222.1 | - | 3 | 984 | |||
AK095763.1 | - | 4 | 656 | |||
AK123341.1 | - | 1 | 819 | |||
AK125100.1 | - | 1 | 1952 | |||
BX354283.2 | - | 7 | 943 | |||
BX641022.1 | - | 1 | 1980 |