Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF280C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017666.4 | NP_060136.1 | 3 | 256 | ||
XM_006724765.3 | XP_006724828.1 | 3 | 207 | |||
GenBank mRNA | AK000102.1 | - | 3 | 179 | ||
AK290373.1 | - | 3 | 207 | |||
AL133204.1 | - | 3 | 191 | |||
AL834333.1 | - | 3 | 187 | |||
BC051728.1 | - | 3 | 207 | 4828435 | ||
DR003059.1 | - | 3 | 255 |