Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKS1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001826.2 | NP_001817.1 | 2 | 263 | ||
NR_024163.1 | 2 | 371 | ||||
GenBank mRNA | AF274941.1 | - | 1 | 185 | ||
AF274943.1 | - | 2 | 191 | |||
AF279897.1 | - | 2 | 185 | |||
AK307066.1 | - | 2 | 319 | |||
AK311769.1 | - | 2 | 224 | |||
BC007751.2 | - | 2 | 206 | 4110344 | ||
BC015629.2 | - | 2 | 185 | 3162564 | ||
BC070319.1 | - | 2 | 224 | 4738841 | ||
BC070320.1 | - | 2 | 224 | 6494754 | ||
BQ883992.1 | - | 2 | 202 | |||
BT007196.1 | - | 2 | 159 | |||
CB161621.1 | - | 2 | 322 | |||
CN347951.1 | - | 2 | 265 | |||
X54941.1 | - | 2 | 168 |