Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193509.1 | NP_001180438.1 | 14 | 1756 | ||
NM_017735.4 | NP_060205.3 | 14 | 2003 | |||
XM_005264416.1 | XP_005264473.1 | 13 | 1867 | |||
GenBank mRNA | AK000279.1 | - | 11 | 1211 | ||
AK000665.1 | - | 9 | 995 | |||
AK027570.1 | - | 14 | 1912 | |||
AK225295.1 | - | 11 | 1211 | |||
AK299196.1 | - | 14 | 1745 | |||
BC001248.1 | - | 13 | 1536 | |||
BC063791.1 | - | 14 | 1961 | 6500951 |