Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193509.1 | NP_001180438.1 | 3, 4 | 418 | ||
NM_017735.4 | NP_060205.3 | 3, 4 | 665 | |||
XM_005264416.1 | XP_005264473.1 | 3, 4 | 652 | |||
XM_011532958.1 | XP_011531260.1 | 3, 4 | 652 | |||
GenBank mRNA | AK027570.1 | - | 3, 4 | 574 | ||
AK299196.1 | - | 3, 4 | 407 | |||
BC001248.1 | - | 2, 3 | 198 | |||
BC063791.1 | - | 3, 4 | 623 | 6500951 |