Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEPDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114120.2 | NP_001107592.1 | 5 | 922 | ||
NM_017779.5 | NP_060249.2 | 5 | 922 | |||
GenBank mRNA | AB281187.1 | - | 5 | 778 | ||
AB281274.1 | - | 5 | 778 | |||
AJ278112.1 | - | 5 | 778 | |||
AK000361.1 | - | 5 | 783 | |||
AK225735.1 | - | 5 | 768 | |||
AK299324.1 | - | 5 | 783 | |||
AL834208.1 | - | 5 | 404 | |||
BC003511.1 | - | 5 | 771 | |||
BC065304.1 | - | 5 | 725 | 6156084 |