Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316690.1 | NP_001303619.1 | 1 | 1142 | ||
NM_017780.3 | NP_060250.2 | 2 | 1460 | |||
XM_011517553.2 | XP_011515855.1 | 2 | 1481 | |||
XM_011517554.2 | XP_011515856.1 | 2 | 1459 | |||
XM_011517555.2 | XP_011515857.1 | 2 | 1481 | |||
XM_011517560.2 | XP_011515862.1 | 2 | 1481 | |||
XM_017013612.1 | XP_016869101.1 | 2 | 1535 | |||
XM_017013613.1 | XP_016869102.1 | 2 | 1481 | |||
GenBank mRNA | AK123158.1 | - | 1 | 251 | ||
BC014681.1 | - | 2 | 1355 | |||
BC053890.1 | - | 2 | 1425 | |||
BC068000.1 | - | 2 | 1445 | |||
BC080627.1 | - | 1 | 1041 | |||
BC110818.1 | - | 2 | 1417 |