Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174081.1 | NP_001167552.1 | 8 | 2377 | ||
NM_001174082.1 | NP_001167553.1 | 7 | 2342 | |||
NM_001174083.1 | NP_001167554.1 | 9 | 2339 | |||
NM_017852.3 | NP_060322.1 | 8 | 2408 | |||
GenBank mRNA | AB209300.1 | - | 7 | 2260 | ||
AF298547.1 | - | 7 | 2259 | |||
AF310106.1 | - | 8 | 2353 | |||
AF464764.1 | - | 8 | 2308 | |||
AK000517.1 | - | 8 | 2353 | |||
AK000784.1 | - | 3 | 528 | |||
AK025952.1 | - | 8 | 2317 | |||
AK223253.1 | - | 8 | 2317 | |||
AK223269.1 | - | 8 | 2317 | |||
AK292681.1 | - | 8 | 2336 | |||
AK302989.1 | - | 9 | 2308 | |||
BC001039.2 | - | 7 | 2288 | 3504594 | ||
BC003592.1 | - | 8 | 2332 | 2821428 | ||
BC039269.1 | - | 8 | 2377 | 5298283 |