Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174081.1 | NP_001167552.1 | 3, 4 | 439 | ||
NM_001174082.1 | NP_001167553.1 | 3, 4 | 470 | |||
NM_001174083.1 | NP_001167554.1 | 4, 5 | 401 | |||
NM_017852.3 | NP_060322.1 | 3, 4 | 470 | |||
GenBank mRNA | AF298547.1 | - | 3, 4 | 387 | ||
AF310106.1 | - | 3, 4 | 415 | |||
AF464764.1 | - | 3, 4 | 370 | |||
AK000517.1 | - | 3, 4 | 415 | |||
AK025952.1 | - | 3, 4 | 379 | |||
AK223253.1 | - | 3, 4 | 379 | |||
AK223269.1 | - | 3, 4 | 379 | |||
AK292681.1 | - | 3, 4 | 407 | |||
AK302989.1 | - | 4, 5 | 370 | |||
BC001039.2 | - | 2, 3 | 350 | 3504594 | ||
BC003592.1 | - | 3, 4 | 394 | 2821428 | ||
BC039269.1 | - | 3, 4 | 439 | 5298283 | ||
DA856893.1 | - | 3, 4 | 439 | |||
DB046128.1 | - | 3, 4 | 470 |