Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136036.2 | NP_001129508.1 | 11 | 1637 | ||
NM_001193328.1 | NP_001180257.1 | 10 | 1189 | |||
NM_017865.3 | NP_060335.2 | 11 | 1421 | |||
XM_011544220.1 | XP_011542522.1 | 11 | 1678 | |||
XM_011544221.1 | XP_011542523.1 | 11 | 1675 | |||
XM_011544222.1 | XP_011542524.1 | 11 | 1293 | |||
XM_011544223.2 | XP_011542525.1 | 10 | 1543 | |||
XM_011544227.1 | XP_011542529.1 | 11 | 1589 | |||
XM_017001698.1 | XP_016857187.1 | 11 | 1290 | |||
XM_017001699.1 | XP_016857188.1 | 10 | 1600 | |||
XM_017001700.1 | XP_016857189.1 | 10 | 1215 | |||
XM_017001701.1 | XP_016857190.1 | 11 | 1405 | |||
XM_017001702.1 | XP_016857191.1 | 10 | 1330 | |||
XM_017001703.1 | XP_016857192.1 | 11 | 1608 | |||
XM_017001704.1 | XP_016857193.1 | 11 | 1565 | |||
XM_017001705.1 | XP_016857194.1 | 11 | 1468 | |||
GenBank mRNA | AB209706.1 | - | 7 | 1475 | ||
AF256175.1 | - | 4 | 738 | |||
AK001070.1 | - | 9 | 837 | |||
AK302187.1 | - | 11 | 1571 | |||
AL133100.1 | - | 9 | 2150 | |||
BC002948.1 | - | 11 | 1377 | 3533691 |