Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194460.2 | NP_919442.1 | 4 | 348 | ||
XM_005259594.1 | XP_005259651.1 | 4 | 428 | |||
GenBank mRNA | AK000559.1 | - | 4 | 299 | ||
AK289616.1 | - | 4 | 348 | |||
BC001442.2 | - | 4 | 321 | 3138961 | ||
BC025374.1 | - | 4 | 275 | 4093350 | ||
BM558139.1 | - | 419 | ||||
BM810439.1 | - | 418 |
Gene Symbol: RNF126P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002818.2 | 1 | 275 |