Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300981.1 | NP_001287910.1 | 6 | 2180 | ||
NM_021130.4 | NP_066953.1 | 5 | 2023 | |||
GenBank mRNA | AK130101.1 | - | 5 | 1980 |