Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP3S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002924.2 | NP_001002924.2 | 7 | 995 | ||
NM_001284.3 | NP_001275.1 | 6 | 959 | |||
NM_001318090.1 | NP_001305019.1 | 4 | 755 | |||
NM_001318091.1 | NP_001305020.1 | 5 | 887 | |||
NM_001318093.1 | NP_001305022.1 | 7 | 1000 | |||
NM_001318094.1 | NP_001305023.1 | 5 | 851 | |||
GenBank mRNA | AK124911.1 | - | 1 | 1660 | ||
AK126876.1 | - | 1 | 4310 | |||
AK311843.1 | - | 6 | 527 | |||
BC000804.1 | - | 6 | 611 | 3452302 | ||
BC012614.1 | - | 7 | 608 | |||
BI600357.1 | - | 6 | 647 | |||
BT009833.1 | - | 6 | 527 | |||
BX394598.2 | - | 5 | 430 | |||
CN411347.1 | - | 5 | 568 | |||
CR450341.1 | - | 6 | 527 | |||
D63643.1 | - | 6 | 569 | |||
DB483797.1 | - | 4 | 441 | |||
U91932.1 | - | 6 | 612 | |||
X99458.1 | - | 6 | 557 |
Gene Symbol: LOC643454 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC157883.1 | - | 1 | 616 | ||
BC171896.1 | - | 1 | 616 |