Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URGCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077663.2 | NP_001071131.1 | 6 | 2902 | ||
NM_001077664.2 | NP_001071132.1 | 6 | 2674 | |||
NM_001290075.1 | NP_001277004.1 | 6 | 2897 | |||
NM_001290076.1 | NP_001277005.1 | 7 | 2796 | |||
NM_017920.4 | NP_060390.3 | 5 | 2875 | |||
GenBank mRNA | AB040940.1 | - | 7 | 3199 | ||
AK000661.1 | - | 1 | 609 | |||
AK026185.1 | - | 1 | 2079 | |||
AK302459.1 | - | 7 | 2796 | |||
AK314339.1 | - | 5 | 2421 | |||
AL832134.1 | - | 4 | 4515 | |||
AL833744.1 | - | 1 | 1003 | |||
AY078404.1 | - | 5 | 2394 | |||
BC018426.1 | - | 5 | 2396 | 4213413 | ||
BX640797.1 | - | 6 | 2437 | |||
BX648505.1 | - | 6 | 2442 | |||
CR749398.1 | - | 6 | 2454 |