Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPLOC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017921.3 | NP_060391.2 | 4 | 582 | ||
XM_011524979.1 | XP_011523281.1 | 4 | 577 | |||
XM_011524980.1 | XP_011523282.1 | 4 | 577 | |||
XM_011524981.1 | XP_011523283.1 | 4 | 577 | |||
XM_011524982.2 | XP_011523284.1 | 4 | 577 | |||
XM_017024830.1 | XP_016880319.1 | 1 | 58 | |||
GenBank mRNA | AB040932.1 | - | 4 | 489 | ||
AK000664.1 | - | 4 | 491 | |||
AK023046.1 | - | 2 | 218 | |||
AK023272.1 | - | 1 | 161 | |||
AK294473.1 | - | 4 | 508 | |||
AL834300.1 | - | 1 | 94 | |||
BC025930.1 | - | 4 | 523 | 5017718 | ||
DC322260.1 | - | 4 | 541 |