Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017925.6 | NP_060395.5 | 22 | 3271 | ||
NR_073201.3 | 22 | 3137 | ||||
XM_006716804.3 | XP_006716867.1 | 23 | 3386 | |||
XM_006716805.3 | XP_006716868.1 | 23 | 3386 | |||
XM_006716807.3 | XP_006716870.1 | 14 | 1911 | |||
XM_011517959.2 | XP_011516261.1 | 23 | 3386 | |||
XM_017014917.1 | XP_016870406.1 | 22 | 3179 | |||
XM_017014918.1 | XP_016870407.1 | 24 | 3586 | |||
XM_017014919.1 | XP_016870408.1 | 24 | 3553 | |||
XM_017014920.1 | XP_016870409.1 | 23 | 3410 | |||
GenBank mRNA | AK025705.1 | - | 2 | 292 | ||
AK125842.1 | - | 16 | 2133 | |||
AK127734.1 | - | 11 | 1547 | |||
AK299842.1 | - | 9 | 1192 | |||
AK302901.1 | - | 4 | 603 | |||
BC144484.1 | - | 19 | 2993 | |||
BC144485.1 | - | 18 | 2369 | |||
CR627367.1 | - | 3 | 477 |