Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033540.2 | NP_284941.2 | 6 | 749 | ||
XM_005247596.3 | XP_005247653.2 | 6 | 707 | |||
XM_011512963.2 | XP_011511265.1 | 3 | 335 | |||
GenBank mRNA | AB043588.1 | - | 5 | 714 | ||
AB043589.1 | - | 6 | 714 | |||
AF329637.1 | - | 5 | 623 | |||
AK000700.1 | - | 6 | 706 | |||
AK314306.1 | - | 6 | 706 | |||
BC020661.1 | - | 6 | 715 | |||
BC040557.1 | - | 6 | 749 | 5270347 | ||
CR627379.1 | - | 3 | 399 | |||
U95822.1 | - | 2 | 212 |