Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193513.1 | NP_001180442.1 | 9 | 640 | ||
NM_001193514.1 | NP_001180443.1 | 8 | 520 | |||
NM_001193515.1 | NP_001180444.1 | 7 | 433 | |||
NM_017964.3 | NP_060434.2 | 8 | 520 | |||
XM_006712048.3 | XP_006712111.1 | 6 | 351 | |||
XM_011532959.2 | XP_011531261.1 | 9 | 617 | |||
XM_011532960.2 | XP_011531262.1 | 8 | 776 | |||
XM_011532961.2 | XP_011531263.1 | 7 | 454 | |||
XM_011532962.2 | XP_011531264.1 | 6 | 605 | |||
XM_011532963.2 | XP_011531265.1 | 9 | 617 | |||
XM_017004462.1 | XP_016859951.1 | 6 | 367 | |||
XM_017004463.1 | XP_016859952.1 | 9 | 617 | |||
XM_017004464.1 | XP_016859953.1 | 9 | 617 | |||
XM_017004465.1 | XP_016859954.1 | 9 | 617 | |||
XM_017004466.1 | XP_016859955.1 | 8 | 512 | |||
GenBank mRNA | AF173387.1 | - | 472 | |||
AK055663.1 | - | 8 | 497 | |||
AK096672.1 | - | 7 | 454 | |||
AK127716.1 | - | 5 | 303 | |||
AK304225.1 | - | 7 | 410 | |||
AL832755.1 | - | 5 | 277 | |||
BC066903.1 | - | 9 | 617 | 4801303 | ||
EF560719.1 | - | 8 | 498 | |||
EF560726.1 | - | 8 | 498 |