Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PACS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018026.3 | NP_060496.2 | 2 | 544 | ||
XM_011545162.1 | XP_011543464.1 | 2 | 104 | |||
XM_011545164.2 | XP_011543466.1 | 2 | 1412 | |||
GenBank mRNA | AK056361.1 | - | 1 | 51 | ||
AK096644.1 | - | 2 | 544 | |||
AK098336.1 | - | 3 | 513 | |||
AY320283.1 | - | 2 | 584 | |||
BC010096.1 | - | 2 | 452 | 3502170 | ||
BC052577.1 | - | 2 | 443 | 4130626 |