Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PACS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018026.3 | NP_060496.2 | 7, 8 | 1121 | ||
XM_011545162.1 | XP_011543464.1 | 7, 8 | 681 | |||
XM_011545164.2 | XP_011543466.1 | 7, 8 | 1989 | |||
GenBank mRNA | AB033001.1 | - | 23 | |||
AK056361.1 | - | 6, 7 | 628 | |||
AK096644.1 | - | 6, 7 | 971 | |||
AY320283.1 | - | 7, 8 | 1161 | |||
BC010096.1 | - | 7, 8 | 1029 | 3502170 | ||
BC052577.1 | - | 7, 8 | 1020 | 4130626 |