Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMD4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318336.1 | NP_001305265.1 | 17 | 2020 | ||
NM_001318337.1 | NP_001305266.1 | 17 | 2260 | |||
NM_001318338.1 | NP_001305267.1 | 7 | 1054 | |||
NM_018027.4 | NP_060497.3 | 18 | 1960 | |||
XM_005252489.2 | XP_005252546.1 | 17 | 2260 | |||
XM_005252490.2 | XP_005252547.1 | 18 | 2056 | |||
XM_005252495.4 | XP_005252552.1 | 4 | 3219 | |||
XM_006717457.2 | XP_006717520.1 | 19 | 2341 | |||
XM_006717458.3 | XP_006717521.1 | 19 | 2341 | |||
XM_006717459.2 | XP_006717522.1 | 18 | 2305 | |||
XM_006717460.2 | XP_006717523.1 | 18 | 2296 | |||
XM_006717464.1 | XP_006717527.1 | 4 | 720 | |||
XM_011519539.1 | XP_011517841.1 | 19 | 2101 | |||
XM_011519540.1 | XP_011517842.1 | 19 | 2341 | |||
XM_011519541.1 | XP_011517843.1 | 19 | 2341 | |||
XM_011519543.2 | XP_011517845.1 | 19 | 2064 | |||
XM_017016393.1 | XP_016871882.1 | 18 | 2305 | |||
XM_017016394.1 | XP_016871883.1 | 18 | 2028 | |||
XM_017016395.1 | XP_016871884.1 | 17 | 1863 | |||
XM_017016396.1 | XP_016871885.1 | 6 | 996 | |||
GenBank mRNA | AB037715.1 | - | 18 | 1960 | ||
AK289693.1 | - | 18 | 1885 | |||
BC151244.1 | - | 18 | 1971 | 8860518 |