Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018052.3 | NP_060522.3 | 11 | 1462 | ||
XM_005256038.3 | XP_005256095.1 | 11 | 1473 | |||
XM_005256041.3 | XP_005256098.2 | 11 | 1473 | |||
XM_011523223.2 | XP_011521525.1 | 11 | 1473 | |||
XM_011523224.2 | XP_011521526.1 | 11 | 1473 | |||
XM_011523225.2 | XP_011521527.1 | 11 | 1473 | |||
XM_017023441.1 | XP_016878930.1 | 11 | 1473 | |||
XM_017023442.1 | XP_016878931.1 | 11 | 1473 | |||
GenBank mRNA | AK025479.1 | - | 5 | 495 | ||
AK056433.1 | - | 11 | 1472 | |||
AK297564.1 | - | 11 | 1247 | |||
AK307766.1 | - | 10 | 1312 | |||
BC007214.1 | - | 6 | 584 | |||
BC125108.1 | - | 11 | 1394 | 40118772 | ||
BC125109.1 | - | 11 | 1394 | 40118777 | ||
CR457343.1 | - | 5 | 430 |