Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160708.1 | NP_001154180.1 | 20 | 2208 | ||
NM_018082.5 | NP_060552.4 | 20 | 2361 | |||
XM_017019621.1 | XP_016875110.1 | 20 | 2326 | |||
GenBank mRNA | AK090636.1 | - | 5 | 433 | ||
AK092031.1 | - | 10 | 1284 | |||
AK122713.1 | - | 20 | 2188 | |||
AK291635.1 | - | 20 | 2205 | |||
AY092084.1 | - | 20 | 2192 | |||
BC046238.1 | - | 20 | 2228 | 4905794 | ||
BX537447.1 | - | 20 | 2309 |