Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145277.1 | NP_001138749.1 | 3 | 364 | ||
NM_001145278.1 | NP_001138750.1 | 3 | 286 | |||
NM_018090.4 | NP_060560.1 | 3 | 364 | |||
XM_011541767.2 | XP_011540069.1 | 4 | 436 | |||
XM_017001721.1 | XP_016857210.1 | 3 | 312 | |||
GenBank mRNA | AK001282.1 | - | 3 | 296 | ||
AK023545.1 | - | 3 | 300 | |||
AK024468.1 | - | 3 | 278 | |||
AK223344.1 | - | 3 | 312 | |||
AK300764.1 | - | 3 | 310 | |||
AK302227.1 | - | 3 | 232 | |||
BC017014.1 | - | 3 | 295 | 3848563 | ||
BC018914.2 | - | 3 | 298 | 4299890 | ||
BM751287.1 | - | 3 | 364 |