Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318716.1 | NP_001305645.1 | 4 | 1692 | ||
NM_001318717.1 | NP_001305646.1 | 4 | 1613 | |||
NM_001318718.1 | NP_001305647.1 | 4 | 1593 | |||
NM_001318719.1 | NP_001305648.1 | 4 | 1571 | |||
NM_001318720.1 | NP_001305649.1 | 4 | 1565 | |||
NM_001318721.1 | NP_001305650.1 | 4 | 1540 | |||
NM_001318722.1 | NP_001305651.1 | 4 | 1561 | |||
NM_001318723.1 | NP_001305652.1 | 4 | 1645 | |||
NM_001318724.1 | NP_001305653.1 | 4 | 1645 | |||
NM_001318725.1 | NP_001305654.1 | 4 | 1548 | |||
NM_016506.6 | NP_057590.3 | 4 | 1649 | |||
NM_018095.5 | NP_060565.4 | 4 | 1544 | |||
XM_017018009.1 | XP_016873498.1 | 4 | 1528 | |||
GenBank mRNA | AK001312.1 | - | 4 | 1498 | ||
AK001749.1 | - | 4 | 1585 | |||
AK091607.1 | - | 3 | 2222 | |||
AK222550.1 | - | 4 | 1495 | |||
AK225062.1 | - | 4 | 1495 | |||
AL713735.1 | - | 3 | 1474 | |||
BC002736.2 | - | 4 | 1586 | 3628290 |