Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271783.1 | NP_001258712.1 | 5 | 1050 | ||
NM_001271784.1 | NP_001258713.1 | 4 | 690 | |||
NM_018099.4 | NP_060569.3 | 5 | 917 | |||
XM_011520747.2 | XP_011519049.1 | 5 | 1320 | |||
XM_011520748.2 | XP_011519050.1 | 5 | 1659 | |||
XM_017019624.1 | XP_016875113.1 | 5 | 1050 | |||
XM_017019625.1 | XP_016875114.1 | 3 | 572 | |||
GenBank mRNA | AK001324.1 | - | 5 | 754 | ||
AK001927.1 | - | 5 | 728 | |||
AK027756.1 | - | 5 | 874 | |||
AK129857.1 | - | 4 | 568 | |||
AK314670.1 | - | 5 | 754 | |||
AL136843.1 | - | 5 | 853 | |||
BC022267.1 | - | 5 | 875 | 4732586 | ||
GQ472211.1 | - | 5 | 875 |