Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018117.11 | NP_060587.8 | 15 | 2141 | ||
XM_005269962.2 | XP_005270019.1 | 15 | 1858 | |||
XM_005269963.2 | XP_005270020.1 | 15 | 1990 | |||
XM_005269964.2 | XP_005270021.1 | 8 | 896 | |||
XM_017016397.1 | XP_016871886.1 | 14 | 1829 | |||
XM_017016398.1 | XP_016871887.1 | 14 | 1759 | |||
XM_017016399.1 | XP_016871888.1 | 14 | 1703 | |||
XM_017016400.1 | XP_016871889.1 | 15 | 2032 | |||
GenBank mRNA | AK001368.1 | - | 10 | 1044 | ||
AK124522.1 | - | 4 | 1790 | |||
AK299524.1 | - | 8 | 761 | |||
BC071564.1 | - | 15 | 1924 | 30346203 |