Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258033.1 | NP_001244962.1 | 17 | 1499 | ||
NM_001258034.1 | NP_001244963.1 | 16 | 1391 | |||
NM_001258035.1 | NP_001244964.1 | 16 | 1425 | |||
NM_001258036.1 | NP_001244965.1 | 17 | 1499 | |||
NM_018119.3 | NP_060589.1 | 17 | 1499 | |||
NR_047581.1 | 17 | 1449 | ||||
XM_006721063.2 | XP_006721126.1 | 17 | 1420 | |||
XM_011545892.1 | XP_011544194.1 | 17 | 1505 | |||
XM_011545893.2 | XP_011544195.1 | 17 | 1508 | |||
XM_017023443.1 | XP_016878932.1 | 17 | 1503 | |||
GenBank mRNA | AB040885.1 | - | 17 | 1419 | ||
AK001371.1 | - | 17 | 1440 | |||
AK023160.1 | - | 17 | 1454 | |||
AK023231.1 | - | 16 | 1377 | |||
AK026645.1 | - | 17 | 1465 | |||
AK222789.1 | - | 17 | 1446 | |||
AK293300.1 | - | 16 | 1270 | |||
AK296813.1 | - | 16 | 1339 | |||
AK300735.1 | - | 17 | 1463 | |||
AY092085.1 | - | 16 | 1298 | |||
BC000285.1 | - | 17 | 1441 | 3356711 |