Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018128.4 | NP_060598.3 | 10 | 2692 | ||
GenBank mRNA | AB037822.1 | - | 10 | 2815 | ||
AK001396.1 | - | 8 | 1334 | |||
AK026565.1 | - | 10 | 1765 | |||
BC019090.2 | - | 9 | 1600 | |||
BC110851.1 | - | 10 | 2270 | 5786818 | ||
BC126110.1 | - | 10 | 2240 | 8991826 |