Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018140.3 | NP_060610.2 | 2 | 233 | ||
XM_005248322.3 | XP_005248379.1 | 2 | 155 | |||
XM_011514063.1 | XP_011512365.1 | 2 | 179 | |||
XM_011514064.2 | XP_011512366.1 | 2 | 286 | |||
XM_017009626.1 | XP_016865115.1 | 2 | 176 | |||
XM_017009627.1 | XP_016865116.1 | 2 | 136 | |||
GenBank mRNA | AB040952.1 | - | 2 | 171 | ||
AK001427.1 | - | 2 | 177 | |||
AK299072.1 | - | 2 | 166 | |||
BC000132.2 | - | 2 | 172 | 2899944 | ||
BC001750.1 | - | 2 | 159 | |||
BG718176.1 | - | 2 | 255 |