Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM222B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077498.2 | NP_001070966.1 | 3 | 352 | ||
NM_001288631.1 | NP_001275560.1 | 4 | 523 | |||
NM_001288632.1 | NP_001275561.1 | 5 | 434 | |||
NM_001288633.1 | NP_001275562.1 | 4 | 428 | |||
NM_001288634.1 | NP_001275563.1 | 3 | 421 | |||
NM_001288635.1 | NP_001275564.1 | 4 | 400 | |||
NM_018182.3 | NP_060652.2 | 4 | 386 | |||
XM_017024831.1 | XP_016880320.1 | 4 | 476 | |||
XM_017024832.1 | XP_016880321.1 | 3 | 741 | |||
XM_017024833.1 | XP_016880322.1 | 3 | 281 | |||
XM_017024834.1 | XP_016880323.1 | 3 | 479 | |||
GenBank mRNA | AK001562.1 | - | 4 | 337 | ||
AK057665.1 | - | 3 | 336 | |||
AK095569.1 | - | 3 | 421 | |||
BC021228.2 | - | 3 | 236 | 4040385 | ||
DA185693.1 | - | 4 | 339 | |||
DA787679.1 | - | 3 | 305 | |||
DB453514.1 | - | 5 | 383 | |||
DC374179.1 | - | 4 | 316 | |||
DC381712.1 | - | 4 | 344 | |||
HY012898.1 | - | 4 | 395 | |||
HY040104.1 | - | 3 | 343 |