Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM222B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077498.2 | NP_001070966.1 | 3 | 331 | ||
NM_001288631.1 | NP_001275560.1 | 4 | 502 | |||
NM_001288632.1 | NP_001275561.1 | 5 | 413 | |||
NM_001288633.1 | NP_001275562.1 | 4 | 407 | |||
NM_001288634.1 | NP_001275563.1 | 3 | 400 | |||
NM_001288635.1 | NP_001275564.1 | 4 | 379 | |||
NM_018182.3 | NP_060652.2 | 4 | 365 | |||
XM_017024831.1 | XP_016880320.1 | 4 | 455 | |||
XM_017024832.1 | XP_016880321.1 | 3 | 720 | |||
XM_017024833.1 | XP_016880322.1 | 3 | 260 | |||
XM_017024834.1 | XP_016880323.1 | 3 | 458 | |||
GenBank mRNA | AK001562.1 | - | 4 | 316 | ||
AK057665.1 | - | 3 | 315 | |||
AK095569.1 | - | 3 | 400 | |||
BC021228.2 | - | 3 | 215 | 4040385 | ||
DA185693.1 | - | 4 | 318 | |||
DA787679.1 | - | 3 | 284 | |||
DB453514.1 | - | 5 | 362 | |||
DC374179.1 | - | 4 | 295 | |||
DC381712.1 | - | 4 | 323 | |||
HY012898.1 | - | 4 | 374 | |||
HY040104.1 | - | 3 | 322 |