Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320047.1 | NP_001306976.1 | 8, 9 | 953 | ||
NM_001320048.1 | NP_001306977.1 | 6, 7 | 708 | |||
NM_018186.3 | NP_060656.2 | 7, 8 | 1256 | |||
XM_005245317.4 | XP_005245374.1 | 7, 8 | 828 | |||
XM_017001722.1 | XP_016857211.1 | 7, 8 | 828 | |||
XM_017001723.1 | XP_016857212.1 | 7, 8 | 828 | |||
XM_017001724.1 | XP_016857213.1 | 5, 6 | 577 | |||
GenBank mRNA | AK001568.1 | - | 6, 7 | 628 | ||
AK096493.1 | - | 8, 9 | 863 | |||
AK127098.1 | - | 11, 12 | 1650 | |||
AK294569.1 | - | 3, 4 | 399 | |||
AK299369.1 | - | 7, 8 | 739 | |||
AK303296.1 | - | 6, 7 | 704 | |||
AL354614.1 | - | 7, 8 | 1256 | |||
BC091516.1 | - | 8, 9 | 867 | 30389801 | ||
BC107169.1 | - | 7, 8 | 744 | 40035375 |