Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320047.1 | NP_001306976.1 | 9, 10 | 1107 | ||
NM_001320048.1 | NP_001306977.1 | 7, 8 | 862 | |||
NM_018186.3 | NP_060656.2 | 8, 9 | 1410 | |||
XM_005245317.4 | XP_005245374.1 | 8, 9 | 982 | |||
XM_017001722.1 | XP_016857211.1 | 8, 9 | 982 | |||
XM_017001723.1 | XP_016857212.1 | 8, 9 | 982 | |||
XM_017001724.1 | XP_016857213.1 | 6, 7 | 731 | |||
GenBank mRNA | AK001568.1 | - | 7, 8 | 782 | ||
AK096493.1 | - | 9, 10 | 1017 | |||
AK127098.1 | - | 12, 13 | 1804 | |||
AK294569.1 | - | 4, 5 | 553 | |||
AK299369.1 | - | 8, 9 | 893 | |||
AK303296.1 | - | 7, 8 | 858 | |||
AL354614.1 | - | 8, 9 | 1410 | |||
BC091516.1 | - | 9, 10 | 1021 | 30389801 | ||
BC107169.1 | - | 8, 9 | 898 | 40035375 |