Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320047.1 | NP_001306976.1 | 10 | 1169 | ||
NM_001320048.1 | NP_001306977.1 | 8 | 924 | |||
NM_018186.3 | NP_060656.2 | 9 | 1472 | |||
XM_005245317.4 | XP_005245374.1 | 9 | 1044 | |||
XM_011509735.2 | XP_011508037.1 | 6 | 654 | |||
XM_017001722.1 | XP_016857211.1 | 9 | 1044 | |||
XM_017001723.1 | XP_016857212.1 | 9 | 1044 | |||
XM_017001724.1 | XP_016857213.1 | 7 | 793 | |||
XM_017001725.1 | XP_016857214.1 | 6 | 654 | |||
GenBank mRNA | AK001568.1 | - | 8 | 844 | ||
AK096493.1 | - | 10 | 1079 | |||
AK127098.1 | - | 13 | 1866 | |||
AK294569.1 | - | 5 | 615 | |||
AK299369.1 | - | 9 | 955 | |||
AK303296.1 | - | 8 | 920 | |||
AL354614.1 | - | 9 | 1472 | |||
BC091516.1 | - | 10 | 1083 | 30389801 | ||
BC107168.1 | - | 8 | 821 | 40035373 | ||
BC107169.1 | - | 9 | 960 | 40035375 |