Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018206.5 | NP_060676.2 | 6 | 643 | ||
XM_005256045.3 | XP_005256102.1 | 5 | 528 | |||
XM_011523227.2 | XP_011521529.1 | 6 | 1373 | |||
GenBank mRNA | AF175265.1 | - | 6 | 601 | ||
AF183418.1 | - | 5 | 552 | |||
AF186382.1 | - | 6 | 597 | |||
AF191298.2 | - | 6 | 571 | |||
AK001614.1 | - | 6 | 591 | |||
AK023650.1 | - | 2 | 154 | |||
AK223218.1 | - | 6 | 562 | |||
AL136888.1 | - | 6 | 591 | |||
BC002414.2 | - | 5 | 550 | 3162255 | ||
BC010362.1 | - | 6 | 564 | 4249949 | ||
BC093036.1 | - | 6 | 589 | 30340379 | ||
BX647213.1 | - | 1 | 933 |